Skip to Content
Merck

Skip To

AB9402

Anti-Serotonin Receptor 1F Antibody

Chemicon®, from rabbit

Synonym(s):

5-HT1F Receptor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 μg

Estimated to ship on19 August 2026fromAreál Kühne+Nagel spol. s r.o.

9 800,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC
Citations:
-

9 800,00 CZK


Estimated to ship on19 August 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HTR1F(3355)

Immunogen

Synthetic peptide from the N-terminal extracellular domain of human Serotonin Receptor 1F. The immunogen sequence has 100% similarity with mouse and rat.

Application

Anti-Serotonin Receptor 1F Antibody detects level of Serotonin Receptor 1F & has been published & validated for use in IH(P).
Optimal working dilutions must be determined by the end user.
Research Category
Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Biochem/physiol Actions

Recognizes Serotonin Receptor 1F [5-HT1F Receptor].

Physical form

Liquid in PBS, pH 7.7 with 0.01% sodium azide.

Preparation Note

Maintain at -70°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters