Skip to Content
Merck
All Photos(1)

Key Documents

AB9088

Sigma-Aldrich

Anti-Polycystin-2 Antibody

serum, Chemicon®

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
CZK 11,800.00

CZK 11,800.00


Estimated to ship onMay 05, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
CZK 11,800.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 11,800.00


Estimated to ship onMay 05, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PKD2(353503)
mouse ... Pkd2(18764)

Specificity

Polycystin-2.

Immunogen

Synthetic peptide from the C-terminal of mouse Polycystin-2.

Application

Anti-Polycystin-2 Antibody is an antibody against Polycystin-2 for use in WB, IC, IH.
Research Category
Metabolism
Research Sub Category
Renal Physiology
Western blot: 1:500 using ECL. The antibody reacts with the 105 kDa Polycystin-2 protein. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody), 1% BSA in 0.1M PBS with 0.05% Tween 20. Preferred gel percentage is 4-12% gradient gel.

Immunocytochemistry: 1:1,000

Immunohistochemistry: 1:1,000 overnight at 2-8°C using a fluorescently labeled secondary antibody. Suggested fixative is 4% paraformaldehyde in 0.1M PBS (one hour). Suggested permeablization method is 0.05% Triton X-100 in dilution buffer. Suggested blocking buffer is 10% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS. Suggested dilution buffer is 1% normal goat serum (or same host as your secondary antibody) and 1% BSA in 0.1M PBS.

Optimal working dilutions must be determined by the end user.

Physical form

Rabbit serum. Liquid.

Storage and Stability

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
POSITIVE CONTROL: adult mouse kidney

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuang Feng et al.
Journal of the American Society of Nephrology : JASN, 28(10), 2973-2984 (2017-06-18)
Autosomal dominant polycystic kidney disease (ADPKD) is caused by inactivating mutations in PKD1 (85%) or PKD2 (15%). The ADPKD proteins encoded by these genes, polycystin-1 (PC1) and polycystin-2 (PC2), form a plasma membrane receptor-ion channel complex. However, the mechanisms controlling
Polycystin-2 expression and function in adult mouse lacrimal acinar cells.
Kaja, S; Hilgenberg, JD; Rybalchenko, V; Medina-Ortiz, WE; Gregg, EV; Koulen, P
Investigative Ophthalmology & Visual Science null
Nathalie Falk et al.
Journal of cell science, 131(16) (2018-07-29)
Pericentrin (Pcnt) is a multifunctional scaffold protein and mutations in the human PCNT gene are associated with several diseases, including ciliopathies. Pcnt plays a crucial role in ciliary development in olfactory receptor neurons, but its function in the photoreceptor-connecting cilium
Shinya Ohata et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 35(31), 11153-11168 (2015-08-08)
Directional beating of ependymal (E) cells' cilia in the walls of the ventricles in the brain is essential for proper CSF flow. E cells display two forms of planar cell polarity (PCP): rotational polarity of individual cilium and translational polarity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service