Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
100 μL | Estimated to ship onMay 27, 2027fromAreál Kühne+Nagel spol. s r.o. | CZK 11,900.00 |
About This Item
CZK 11,900.00
Estimated to ship onMay 27, 2027Details
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
serum
antibody product type
primary antibodies
clone
polyclonal
species reactivity
mouse, human, rat
manufacturer/tradename
Chemicon®
technique(s)
western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... IMMT(10989)
General description
Application
Western Blot: 1:5000
Cell Structure
Cytoskeleton
Physical form
Preparation Note
Analysis Note
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form serum | antibody form affinity isolated antibody | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| biological source rabbit | biological source rabbit | biological source mouse | biological source mouse |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| Quality Level 100 | Quality Level 200 | Quality Level - | Quality Level 100 |
| clone polyclonal | clone polyclonal | clone 1A8, monoclonal | clone 3C9, monoclonal |
| species reactivity mouse, human, rat | species reactivity human, mouse, rat | species reactivity human | species reactivity human |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



