Skip to Content
Merck

Skip To

AB5603A4

Anti-Sox2, Alexa Fluor 488 Conjugate Antibody

from rabbit, ALEXA FLUOR 488

Synonym(s):

Transcription factor SOX-2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship on10 July 2026fromAreál Kühne+Nagel spol. s r.o.

14 300,00 CZK

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
ALEXA FLUOR 488
Clone:
polyclonal
Application:
ICC
Citations:
2

14 300,00 CZK


Estimated to ship on10 July 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

ALEXA FLUOR 488

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

species reactivity (predicted by homology)

mouse (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SOX2(6657)

General description

SOX2 is also known as SRY related HMG BOX gene 2. All SOX proteins have a single HMG box and bind linear DNA in a sequence-specific manner, resulting in the bending of DNA through large angles. Bending causes the DNA helix to open for some distance, which may affect binding and interactions of other transcription factors. SOX1, SOX2 and SOX3 show the closest homology to SRY. They share maximum homology within the HMG domain and are expressed mainly in the developing nervous system of the mouse. These genes share significant homology outside the HMG box also and are highly conserved throughout their evolution.
The unconjugated antibody (AB5603) tested on western blot detects Sox2 in whole cell or nuclear extracts as a protein band at 34 kDa

Immunogen

Synthetic peptide from human Sox2.

Application

Detect Sox2 using this Anti-Sox2, Alexa Fluor 488 Conjugate Antibody validated for use in ICC.
Research Category
Neuroscience
Research Sub Category
Transcription Factors

Physical form

Affinity purified
Purified rabbit polyclonal conjugated to Alexa Fluor 488 in PBS with 0.1% sodium azide and 15 mg/mL BSA.

Preparation Note

Maintain refrigerated at 2-8 °C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Undifferentiated ReNcell VM neural progenitor cells
Evaluated by Immunocytochemistry in undifferentiated ReNcell VM neural progenitor cells.

Immunocytochemistry Analysis: A 1:100 dilution of this antibody detected Sox2 in undifferentiated ReNcell VM neural progenitor cells.

Legal Information

ALEXA FLUOR is a trademark of Life Technologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB5603SAB5700644S9072
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100, 300

Quality Level

100

Quality Level

200

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

ALEXA FLUOR 488

conjugate

-

conjugate

-

conjugate

unconjugated

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

UniProt accession no.

P48431

UniProt accession no.

P48431

UniProt accession no.

P48431

UniProt accession no.

P48431


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters