Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
50 μg | Estimated to ship onJune 12, 2026fromAreál Kühne+Nagel spol. s r.o. | CZK 12,800.00 |
About This Item
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human
manufacturer/tradename
Chemicon®
technique(s)
immunohistochemistry: suitable (paraffin)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... CXCR3(2833)
General description
Immunogen
Application
Optimal working dilutions must be determined by the end user.
Inflammation & Immunology
Cytokines & Cytokine Receptors
Biochem/physiol Actions
Physical form
Preparation Note
Other Notes
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| biological source rabbit | biological source mouse | biological source rabbit | biological source mouse |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| antibody form affinity purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
| Quality Level 100 | Quality Level 200 | Quality Level 100 | Quality Level 100 |
| Gene Information human ... CXCR3(2833) | Gene Information human ... CXCR3(2833) | Gene Information human ... CX3CR1(1524) | Gene Information human ... CXCR4(7852) |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



