Skip to Content
Merck

Skip To

AB3271-I

Anti-ASCIZ Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

ATM interactor, ATM/ATR-substrate CHK2-interacting zinc finger protein, ASCIZ, Zinc finger protein 822

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg

Estimated to ship onJune 09, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 9,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
1

CZK 9,800.00


Estimated to ship onJune 09, 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATMIN(23300)

General description

ASCIZ is a transcription factor that interacts with the potent kinase ATM and regulates its activity and stability. ATM is best known for its role as an apical activator of the DNA damage response in the face of DNA double-strand breaks (DSBs). Following induction of DSBs, ATM mobilizes one of the most extensive signaling networks that responds to specific stimuli and modifies directly or indirectly a broad range of targets. ASCIZ plays a crucial role in cell survival and RAD51 foci formation in response to methylating DNA damage as well. ASCIZ is also critical in development. Knockouts are lethal at E16 and show severe organ developmental defects, including a complete absence of lungs similar to mutants in Wnt2-2b/ß-catenin and FGF10/FGFR2b signaling pathways. Thus, ASCIZ has dual functions as an efficiency factor for DNA base damage repair as well as a key transcriptional regulator of early lung development. ASCIZ is localized to the nucleus in discrete foci during G1 and is ubiquitously expressed in normal and cancerous tissues.
~100/120 kDa observed. Uncharacterized band(s) may appear in some lysates.

Immunogen

Epitope: Required for formation of RAD51 foci
KLH-conjugated linear peptide corresponding to human ASCIZ required for formation of RAD51 foci.

Application

Anti-ASCIZ is an antibody against ASCIZ for use in Western Blotting.

Analysis Note

Evaluated by Western Blotting in human skeletal muscle tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected ASCIZ in 10 µg of human skeletal muscle tissue lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB4340ABE1331SAB1402696
conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

mouse

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

2C10, monoclonal

Gene Information

human ... ATMIN(23300)

Gene Information

-

Gene Information

human ... IKZF4(64375)

Gene Information

human ... ATM(472)


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters