Skip to Content
Merck
All Photos(1)

Key Documents

AB2953

Sigma-Aldrich

Anti-ATG3 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

ATG3 autophagy related 3 homolog (S. cerevisiae), APG3 autophagy 3-like (S. cerevisiae), autophagy-related protein 3, autophagy Apg3p/Aut1p-like, APG3 autophagy 3-like, Protein PC3-96

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
CZK 12,200.00

CZK 12,200.00


Estimated to ship onApril 01, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μG
CZK 12,200.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

CZK 12,200.00


Estimated to ship onApril 01, 2025Details


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ATG3(64422)

General description

ATG3 is thought to be an autophagy-related ubiquitin carrier protein (E2)-like enzyme that is the go between for the conjugation on ATG8 and phosphatidylethanolamine (PE). This compound is necessary for autophagy, which is a process in which a cell is degraded by its own lysosomal process.

Specificity

Other homologies: Ape, rhesus macaque, equine, porcine, and bovine (99% sequence homology). Mouse (98% sequence homology). Rat (97% sequence homology).

Immunogen

GST-tagged recombinant protein corresponding to human ATG3.

Application

Anti-ATG3 Antibody detects level of ATG3 & has been published & validated for use in WB.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blot in HL-60 cell lysate.

Western Blot Analysis: 0.1 µg/mL of this antibody detected ATG3 in 10 µg of HL-60 cell lysate.

Target description

~37 kDa observed

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
HL-60 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service