Skip to Content
Merck

Skip To

AB15690

Anti-Orexin A Antibody

Chemicon®, from chicken

Synonym(s):

Hypocretin 1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μL

Estimated to ship onJune 10, 2026fromAreál Kühne+Nagel spol. s r.o.

CZK 11,800.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ELISA
Citations:
-

CZK 11,800.00


Estimated to ship onJune 10, 2026Details


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

chicken

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPOX(5498)

Immunogen

Synthetic peptide from the N-terminus of human Orexin A.

Application

Anti-Orexin A Antibody detects level of Orexin A & has been published & validated for use in ELISA.
ELISA: 1:200-1:1,000 when using as the coating antibody and 1:2,000-1:5,000 when using as the detection antibody in a sandwich ELISA.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
CNS Control of Metabolism

Circadian Rhythm & Sleep

Biochem/physiol Actions

Orexin A

Physical form

ImmunoAffinity Purified
Liquid in PBS with 0.02% sodium azide.

Preparation Note

Maintain at -20°C in undiluted aliquots for up to 6 months after date of receipt. Avoid repeated freeze/thaw cycles.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AB3098AB3096AB3092
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

biological source

chicken

biological source

rabbit

biological source

rabbit

biological source

rabbit

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

shipped in

wet ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

Gene Information

human ... PPOX(5498)

Gene Information

human ... PPOX(5498)

Gene Information

human ... PPOX(5498)

Gene Information

human ... HCRTR1(3061)


Still not finding the right product?

Explore all of our products under Anti-Orexin A Antibody

or

Try our Product Selector Tool to narrow your options


wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

12 - Non Combustible Liquids



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters