Skip to Content
Merck
All Photos(1)

Documents

AB1498P

Sigma-Aldrich

Anti-Stem Cell Factor Antibody

Chemicon®, from rabbit

Synonym(s):

SCF, c-kit Ligand, Mast Cell Growth Factor, MGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
neutralization: suitable
western blot: suitable

input

sample type hematopoietic stem cell(s)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... KITLG(4254)

General description

Stem Cell Factor (SCF), also known as c-Kit ligand (KL), steel factor (SLF) and mast cell growth factor (MGF), is a 30 kDa glycoprotein with broad activities on various tissues, including hematopoietic cells, pigment cells, and primordial germ cells. SCF is secreted by endothelial cells, fibroblasts, and bone marrow stromal cells as a membrane-bound form which may be cleaved to release the soluble form. Both forms are active in promoting colony formation from murine bone marrow cells, but membrane-bound SCF is more effective in promoting hematopoieses in vivo, suggesting a role in cellular interactions between hematopoietic and stromal cells. The soluble form is thought to exist in solution as a noncovalently linked dimer. SCF is structurally related to M-CSF (CSF-1) and Flt-3/Flk-2 Ligand (FL) with all three sharing a similar size, existence of transmembrane and soluble forms, four conserved cysteines, and alternative splicing exon locations, but they share little sequence homology. SCF alone is a modest colony stimulating factor. However, in the presence of other cytokines such as EPO, TPO, GM-CSF, G-CSF, M-CSF, IL-3, and IL-7, SCF is a potent costimulant that works synergistically to increase the size of myeloid, erythroid or lymphoid lineage colonies without influencing the lineage differentiation of the progenitors.

Specificity

Reacts with mouse SCF; SCF is highly conserved cross reactivity with rat, human and other species SCF is expected but untested.

Immunogen

Highly pure (>98%) recombinant mouse SCF

Application

Detect Stem Cell Factor using this Anti-Stem Cell Factor Antibody validated for use in ELISA, WB, NEUT.
ELISA: To detect mSCF by direct ELISA (using 100 μL/well antibody solution) a concentration of at lease 0.5 μg/mL of this antibody is required. Should allow detection of 0.2 - 0.4 ng/well of recombinant mSCF.

Neutralization: For ½ maximal inhibition (ND50) of the bioogical activity of mSCF (20 ng/mL), a concentration of 0.64 - 0.97 μg/mL is required.

Western blot: 0.1 - 0.2 μg/mL, this dilution should offer a detection limit of 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.

Optimal working dilutions must be determined by the end user.
Research Category
Stem Cell Research
Research Sub Category
Hematopoietic Stem Cells

Target description

30 kDa

Physical form

Affinity purified immunoglobulin. Lyophilized from 0.5X PBS pH 7.2. Reconstitute with sterile distilled water.

Storage and Stability

The lyophilized antibody is stable at room temperature for at least one month and for greater than a year when kept at -20ºC. When reconstituted in sterile water to a concentration of = 0.2 mg/ml the antibody is stable for at least six weeks at 2-4ºC.

Analysis Note

Control
Human mast cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service