Skip to Content
Merck

Skip To

9.43441

Stainless steel drum, 190 l

2 in. opening, 3/4 in. opening

Synonym(s):

Drum VA 190 l

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.51
UNSPSC Code:
24112108
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


fitting

2 in. opening, 3/4 in. opening

General description

We offer a big variety of packaging materials like empty bottles and containers for your own preparations and mixtures of our reagents.
with 2" and 3/4" threaded opening




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. 1. is 9.43441.1012 is for 190 L 2. is this empty drum food Grade. user will use this drum to store their product, neutral alcohol neutral alcohol (45% ABV). 3. do you have stock for 6 x 190 L now? 4. please inform the dimension, include weight ? thanks

    1 answer
    1. Product 9434411012 is a 10 liter drum and 9434411028 is a 25 liter drum. These drums are not tested or suitable for any food grade applications. They are suitable for contents that are intended for research use only.

      The dimensions of the 10L stainless steel drum is as follows:
      Height: 35 cm
      Diameter: 24 cm
      Volume: 12 L
      Weight: 1.9 kg

      The dimensions of the 25L stainless steel drum is as follows:
      Height: 52 cm
      Diameter: 29 cm
      Volume: 28 L
      Weight: 3.8 kg

      For product availability please contact customer service. Please navigate to the link https://www.sigmaaldrich.com/support/customer-support, click on "Products Pricing & Availability" under the Products Section with all the required information so that a member of the Customer Service team can reach out to assist further. Thank you.

      Helpful?

Reviews

No rating value

Active Filters