Skip to Content
Merck
All Photos(1)

Documents

9035-HK

Sigma-Aldrich

Glucagon Like Peptide 1 (GLP-1)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.13

conjugate

125Iodine

Quality Level

label

125Iodine labeled

NCBI accession no.

UniProt accession no.

Gene Information

human ... GCG(2641)

General description

Radioactivity: <1.5 μCi
These iodinated hormonos are HPLC purified and prepared fresh monthly.

Application

Research Category
All
appropriate label hydrating buffer if kit replacement or assay buffer of choice

Storage and Stability

2–8 °C

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service