Skip to Content
Merck

8.56188

H-His(Trt)-Trityl NovaPEG resin

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
51171641
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

granular

manufacturer/tradename

Novabiochem®

storage temp.

2-8°C

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_c

Not applicable

flash_point_f

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
856188000104027269165956
856188000504027269165963