Skip to Content
Merck

Skip To

8.56067

H-Arg(Pbf)-2-ClTrt resin

Novabiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Please contact Customer Service for Availability
CZK 6,270.00

About This Item

NACRES:
NA.22
UNSPSC Code:
12352209

CZK 6,270.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

beads

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

storage temp.

2-8°C

General description

Pre-loaded resin for synthesis of protected peptide acids containing a C-terminal arginine amino-acid residue by Fmoc SPPS. Cleavage of the protected peptide is effected by treatment with 1% TFA in DCM or 20% TFE in DCM

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to yellow to beige
Appearance of substance (visual): beads
Loading (determined from the substitution of the Fmoc-Leu loaded resin): 0.30 - 0.90 mmol/g
Swelling Volume (in DMF): lot specific result
Identity (of the substitution): passes test
The polymer matrix is copoly (styrene - 1% DVB), 200 - 400 mesh.

Other Notes

Replaces: 04-12-2823

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.561428.560598.56062
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

form

beads

form

beads

form

beads

form

solid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters