Skip to Content
Merck

Skip To

8.55152

SEA-PS resin

Novabiochem®

Synonym(s):

bis(2-Sulfanylethyl)aminotrityl polystyrene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
CZK 7,410.00
5 g
Please contact Customer Service for Availability
CZK 23,900.00

About This Item

NACRES:
NA.22
UNSPSC Code:
12162002

CZK 7,410.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

amine

storage temp.

2-8°C

General description

A novel resin for preparation peptide thioesters by Fmoc SPPS.

Associated Protocols and Technical Articles
Protocols for Loading of Peptide Synthesis Resins

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
549428M1773M9773
reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

reaction suitability

reaction type: Boc solid-phase peptide synthesis

reaction suitability

-

reaction suitability

-

functional group

amine

functional group

-

functional group

-

functional group

-

product line

Novabiochem®

product line

-

product line

-

product line

-

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

-

application(s)

peptide synthesis

application(s)

-

application(s)

-

application(s)

-

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters