Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 g | Please contact Customer Service for Availability | 9 070,00 CZK 6 802,50 CZK |
About This Item
Empirical Formula (Hill Notation):
C14H25NO6
CAS Number:
Molecular Weight:
303.35
UNSPSC Code:
12352209
NACRES:
NA.22
MDL number:
6 802,50 CZK
List Price9 070,00 CZKSave 25%Web-Only Promotion
Please contact Customer Service for Availability
Quality Segment
product line
Novabiochem®
assay
≥98% (TLC)
form
powder
reaction suitability
reaction type: Boc solid-phase peptide synthesis
manufacturer/tradename
Novabiochem®
mp
102-105 °C
application(s)
peptide synthesis
functional group
carboxylic acid
storage temp.
2-30°C
SMILES string
N([C@@H](CCC(=O)OC(C)(C)C)C(=O)[O-])C(=O)OC(C)(C)C
InChI
1S/C14H25NO6/c1-13(2,3)20-10(16)8-7-9(11(17)18)15-12(19)21-14(4,5)6/h9H,7-8H2,1-6H3,(H,15,19)(H,17,18)/p-1/t9-/m0/s1
InChI key
YGSRAYJBEREVRB-VIFPVBQESA-M
General description
Building block of introduction of N-terminal glutamic acid amino-acid residues by Fmoc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS
Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Boc SPPS
Analysis Note
Colour (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -10.5 - -8.5 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble
To see the solvent systems used for TLC of Novabiochem® products please click here.
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in methanol): -10.5 - -8.5 °
Purity (TLC(011A)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Solubility (1 mmole in 2 ml DMF): clearly soluble
To see the solvent systems used for TLC of Novabiochem® products please click here.
Other Notes
Replaces: 04-12-0051
Legal Information
Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Boc-Glu(OtBu)-OH
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.