Skip to Content
Merck

Skip To

8.52245

Fmoc-D-Thr(PO(OBzl)OH)-OH

Novabiochem®

Synonym(s):

Fmoc-D-Thr(PO(OBzl)OH)-OH, N-α-Fmoc-O-benzyl-D-phosphothreonine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C26H26NO8P
CAS Number:
Molecular Weight:
511.46
UNSPSC Code:
12352209
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

Novabiochem®

assay

≥90.0% (acidimetric), ≥95.0% (HPLC), ≥97% (TLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

15-25°C

General description

An excellent building block for the preparation of D-phosphothreonine-containing peptides by Fmoc SPPS. This derivative can be introduced using standard activation methods, such as PyBOP® and TBTU. The monoprotected phosphoserine residue once incorporated is stable to piperidine. Using this reagent, even peptides containing multiple phosphorylation sites can be prepared efficiently by standard Fmoc SPPS methods.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS
Fmoc SPPS of Peptides containing Post-Translational Modifications

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in DMF): +3.5 - +6.5 °
Purity (TLC(CMA1)): ≥ 97 %
Assay (HPLC, area%): ≥ 95.0 % (a/a)
Solubility (1 mmole in 2 ml DMF): clearly soluble
Assay (acidimetric): ≥ 90.0 %
Water (K. F.): ≤ 2.00 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-13-1079

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany
PyBOP is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters