Skip to Content
Merck

Skip To

8.52207

Fmoc-Trp-OH

Novabiochem®

Synonym(s):

Fmoc-Trp-OH, N-α-Fmoc-L-tryptophan

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Please contact Customer Service for Availability
CZK 582.00
25 g
Please contact Customer Service for Availability
CZK 1,520.00
100 g
Please contact Customer Service for Availability
CZK 4,180.00
Please contact Customer Service for Availability
CZK 33,700.00

About This Item

Empirical Formula (Hill Notation):
C26H22N2O4
CAS Number:
Molecular Weight:
426.46
UNSPSC Code:
12352209
EC Index Number:
252-706-5
NACRES:
NA.22
MDL number:

CZK 582.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥96.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-30°C

SMILES string

[nH]1c2c(c(c1)C[C@H](NC(=O)OCC3c4c(cccc4)c5c3cccc5)C(=O)[O-])cccc2

InChI

1S/C26H22N2O4/c29-25(30)24(13-16-14-27-23-12-6-5-7-17(16)23)28-26(31)32-15-22-20-10-3-1-8-18(20)19-9-2-4-11-21(19)22/h1-12,14,22,24,27H,13,15H2,(H,28,31)(H,29,30)/p-1/t24-/m0/s1

InChI key

MGHMWKZOLAAOTD-DEOSSOPVSA-M

General description

Building block for introduction of typtophan by Fmoc SPPS. Superceded by Fmoc-Trp(Boc)-OH.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Enantiomeric purity: ≥ 99.0 % (a/a)
Purity (TLC(011C)): ≥ 98 %
Purity (TLC(0811)): ≥ 98 %
Assay (HPLC, area%): ≥ 98.0 % (a/a)
Solubility (25 mmole in 50 ml DMF): clearly soluble
Assay (acidimetric): ≥ 96.0 %
Water (K. F.): ≤ 1.0 %

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1035

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.521508.521648.52131
form

powder

form

powder

form

powder

form

powder

assay

≥96.0% (acidimetric), ≥98.0% (HPLC), ≥98% (TLC)

assay

≥96.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

assay

≥85.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

assay

≥95.0% (HPLC)

functional group

Fmoc

functional group

Fmoc

functional group

Boc, Fmoc

functional group

Boc, Fmoc

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

15-25°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under Fmoc-Trp-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters