Skip to Content
Merck

Skip To

8.52129

Fmoc-Lys(Boc)-OPfp

Novabiochem®

Synonym(s):

Fmoc-Lys(Boc)-OPfp, N-α-Fmoc-N-ε-t.-Boc-L-lysine pentafluorophenyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C32H31F5N2O6
CAS Number:
Molecular Weight:
634.59
MDL number:
UNSPSC Code:
12352209
NACRES:
NA.22
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

Novabiochem®

assay

≥97.0% (HPLC)

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

manufacturer/tradename

Novabiochem®

mp

135-145 °C

application(s)

peptide synthesis

functional group

amine

storage temp.

15-25°C

SMILES string

Fc1c(c(c(c(c1F)F)OC(=O)[C@@H](NC(=O)OCC2c3c(cccc3)c4c2cccc4)CCCCNC(=O)OC(C)(C)C)F)F

InChI

1S/C32H31F5N2O6/c1-32(2,3)45-30(41)38-15-9-8-14-22(29(40)44-28-26(36)24(34)23(33)25(35)27(28)37)39-31(42)43-16-21-19-12-6-4-10-17(19)18-11-5-7-13-20(18)21/h4-7,10-13,21-22H,8-9,14-16H2,1-3H3,(H,38,41)(H,39,42)/t22-/m0/s1

InChI key

HLNVSYQQDWNJRI-QFIPXVFZSA-N

General description

Pre-formed pentafluorophenyl ester for coupling of lysine amino-acid residues by Fmoc SPPS. Use enables bromophenol blue monitoring of amide bond formation.

Associated Protocols and Technical Articles
Cleavage and Deprotection Protocols for Fmoc SPPS

Analysis Note

Color (visual): white to slight yellow to beige
Appearance of substance (visual): powder
Identity (IR): passes test
Optical rotation α 25/D (c=1 in chloroform): -16.0 - -13.0 °
Purity (TLC (0004)): <2% DCU
Assay (HPLC, area%): ≥ 97.0% (a/a)
Solubility (0,5 mmole in 3 ml DMF): clearly soluble

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 04-12-1511

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.520128.521288.52146
form

powder

form

powder

form

powder

form

powder

assay

≥97.0% (HPLC)

assay

≥98% (TLC), ≥98.0% (acidimetric), ≥99.0% (HPLC)

assay

≥98.0% (HPLC)

assay

≥97.0% (acidimetric), ≥98% (TLC), ≥98.0% (HPLC)

functional group

amine

functional group

Boc, amine

functional group

Fmoc

functional group

amine

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

15-25°C

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

15-25°C

mp

135-145 °C

mp

123-130 °C

mp

150-160 °C

mp

120-130 °C


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters