Skip to Content
Merck

Skip To

8.51035

Fmoc-NH-(PEG)3-COOH (16 atoms)

≥98% (TLC), for peptide synthesis, Novabiochem®

Synonym(s):

Fmoc-NH-(PEG)₃-COOH (16 atoms), Fmoc-15-amino-4,7,10,13-tetraoxapentadecanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
CZK 15,200.00

About This Item

Empirical Formula (Hill Notation):
C26H33NO8
CAS Number:
Molecular Weight:
487.54
UNSPSC Code:
12352106
NACRES:
NA.22
MDL number:

CZK 15,200.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Fmoc-NH-(PEG)3-COOH (16 atoms), Novabiochem®

Quality Segment

product line

Novabiochem®

assay

≥97.5% (HPLC), ≥98% (TLC)

form

oil

reaction suitability

reaction type: Pegylations

manufacturer/tradename

Novabiochem®

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

N(CCOCCOCCOCCOCCC(=O)O)C(=O)OCC1c2c(cccc2)c3c1cccc3

InChI

1S/C26H33NO8/c28-25(29)9-11-31-13-15-33-17-18-34-16-14-32-12-10-27-26(30)35-19-24-22-7-3-1-5-20(22)21-6-2-4-8-23(21)24/h1-8,24H,9-19H2,(H,27,30)(H,28,29)

InChI key

NUHRPLKTAAVHCZ-UHFFFAOYSA-N

General description

16-Atom long aminoacid PEG spacer. Can be introduced using standard Fmoc SPPS protocols

Application

Fmoc-NH-(PEG)₃-COOH (16 atoms) Novabiochem®. CAS 557756-85-1, molar mass 487.55 g/mol.

Analysis Note

Colour (visual): colorless to slight yellow to beige
Appearance of substance (visual): oil
Identity (LC-MS): 486.2 - 488.2
Purity (TLC(011A)): ≥ 98 %
Assay (HPLC, area%): ≥ 97.5 % (a/a)

To see the solvent systems used for TLC of Novabiochem® products please click here.

Other Notes

Replaces: 01-63-0199

Legal Information

Novabiochem is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.510368.510348.51038
form

oil

form

oil

form

solid

form

oil

assay

≥97.5% (HPLC), ≥98% (TLC)

assay

≥97% (TLC), ≥97.0% (HPLC)

assay

≥98% (TLC), ≥99.0% (HPLC)

assay

≥93.0% (HPLC), ≥95% (TLC)

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

product line

Novabiochem®

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters