Skip to Content
Merck

Skip To

8.45017

N-Ethyldiisopropylamine

Msynthplus

Synonym(s):

N-Ethyldiisopropylamine, N,N-Diisopropylethylamine

Sign In to View Organizational & Contract Pricing.

About This Item

Linear Formula:
[(CH3)2CH]2NCH2CH3
CAS Number:
Molecular Weight:
129.24
EC Index Number:
230-392-0
UNSPSC Code:
12352116
MDL number:
Assay:
≥99.5% (GC)
Bp:
127 °C/1013 hPa
Vapor pressure:
14 hPa ( 20 °C)


vapor pressure

14 hPa ( 20 °C)

assay

≥99.5% (GC)

form

liquid

autoignition temp.

240 °C

potency

200-500 mg/kg LD50, oral (Rat)

expl. lim.

0.7-6.3 % (v/v)

dilution

(for synthesis)

pH

12.3 (20 °C in H2O, as an emulsion)

kinematic viscosity

0.88 cSt(20 °C)

bp

127 °C/1013 hPa

transition temp

flash point 9.5 °C

density

0.76 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H19N/c1-6-9(7(2)3)8(4)5/h7-8H,6H2,1-5H3

InChI key

JGFZNNIVVJXRND-UHFFFAOYSA-N

Analysis Note

Colour (visual): colourless to yellowish
Appearance (visual): clear
Assay (GC, area%): ≥ 99.5 % (a/a)
Density (d 20 °C/ 4 °C): 0.755 - 0.758
Water (K. F.): ≤ 0.20 %
Identity (IR): passes test
1,1,1-Trichloroethane (HS-GC): lot specific result
Benzene (HS-GC): lot specific result
1,1-Dichlorethene: not measurable
Carbon tetrachloride: not measurable
1,2-Dichlorethane: not measurable

Legal Information

MSYNTH is a trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under N-Ethyldiisopropylamine


signalword

Danger

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_c

9.5 °C

flash_point_f

49.1 °F

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Flam. Liq. 2 - STOT SE 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters