Skip to Content
Merck

Skip To

8.44010

Decamethyltetrasiloxane

for synthesis

Synonym(s):

Decamethyltetrasiloxane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 L
Please contact Customer Service for Availability
CZK 21,900.00

About This Item

CAS Number:
Molecular Weight:
310.69
UNSPSC Code:
12352103
EC Index Number:
205-491-7
NACRES:
NA.22
MDL number:

CZK 21,900.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

73 Pa ( 25 °C)

Quality Segment

form

liquid

potency

>2000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rat)

kinematic viscosity

1.3 cSt(20 °C)

mp

-76 °C

transition temp

flash point 62 °C

solubility

<0.1 mg/L

density

0.85 g/cm3 at 25 °C

storage temp.

2-30°C

SMILES string

[Si](O[Si](O[Si](C)(C)C)(C)C)(O[Si](C)(C)C)(C)C

InChI

1S/C10H30O3Si4/c1-14(2,3)11-16(7,8)13-17(9,10)12-15(4,5)6/h1-10H3

InChI key

YFCGDEUVHLPRCZ-UHFFFAOYSA-N

General description

for synthesis

Application

Decamethyltetrasiloxane for synthesis. CAS 141-62-8, molar mass 310.69 g/mol.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
2356791.010071.00163
form

liquid

form

liquid

form

solid

form

solid

vapor pressure

73 Pa ( 25 °C)

vapor pressure

-

vapor pressure

-

vapor pressure

-

solubility

<0.1 mg/L

solubility

-

solubility

166.5 g/L

solubility

36 g/L

transition temp

flash point 62 °C

transition temp

-

transition temp

-

transition temp

-

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

mp

-76 °C

mp

−68 °C (lit.)

mp

295-297 °C (decomposition)

mp

450 °C


Still not finding the right product?

Explore all of our products under Decamethyltetrasiloxane


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

10 - Combustible liquids

wgk

WGK 3

flash_point_f

143.6 °F - closed cup

flash_point_c

62 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters