Skip to Content
Merck
All Photos(1)

Key Documents

8.43970

Sigma-Aldrich

N-Ethyltoluene-4-sulfonamide

for synthesis

Synonym(s):

N-Ethyltoluene-4-sulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
CZK 5,830.00

CZK 5,830.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
CZK 5,830.00

About This Item

Empirical Formula (Hill Notation):
C9H13NO2S
CAS Number:
Molecular Weight:
199.27
MDL number:
UNSPSC Code:
12352100
EC Index Number:
201-275-1
NACRES:
NA.22

CZK 5,830.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

solid

mp

63-65 °C

transition temp

flash point 127 °C

density

1.26 g/cm3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)(NCC)c1ccc(cc1)C

InChI

1S/C9H13NO2S/c1-3-10-13(11,12)9-6-4-8(2)5-7-9/h4-7,10H,3H2,1-2H3

InChI key

OHPZPBNDOVQJMH-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service