Skip to Content
Merck
All Photos(1)

Documents

8.43910

Sigma-Aldrich

[2-(2-Methoxyethoxy)ethoxy] acetic acid

for synthesis

Synonym(s):

[2-(2-Methoxyethoxy)ethoxy] acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14O5
CAS Number:
Molecular Weight:
178.18
MDL number:
UNSPSC Code:
12352106
EC Index Number:
240-162-1
NACRES:
NA.22

Quality Level

pH

1.8 (100 g/L in H2O)

bp

140 °C/2 mbar

transition temp

flash point >160 °C

density

1.157 g/cm3

storage temp.

2-30°C

InChI

1S/C7H14O5/c1-10-2-3-11-4-5-12-6-7(8)9/h2-6H2,1H3,(H,8,9)

InChI key

YHBWXWLDOKIVCJ-UHFFFAOYSA-N

Application


  • Dextromethorphan Impurity C in biochemical analysis: A method utilizing pipette tip solid-phase extraction and gas chromatography-mass spectrometry was developed for the determination of dextromethorphan in human plasma, demonstrating a precise approach to detect and analyze impurities like Dextromethorphan Impurity C in pharmaceutical compounds (Hasegawa et al., 2011).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service