Skip to Content
Merck

Skip To

8.43791

1,2,4-Trihydroxybenzene

for synthesis

Synonym(s):

1,2,4-Trihydroxybenzene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 g
Please contact Customer Service for Availability
CZK 3,350.00
25 g
Please contact Customer Service for Availability
CZK 11,500.00

About This Item

Empirical Formula (Hill Notation):
C6H6O3
CAS Number:
Molecular Weight:
126.11
UNSPSC Code:
12352104
EC Index Number:
208-575-1
NACRES:
NA.22
MDL number:

CZK 3,350.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (HPLC)

form

solid

mp

137-140 °C

density

1.45 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

Oc1c(ccc(c1)O)O

InChI

1S/C6H6O3/c7-4-1-2-5(8)6(9)3-4/h1-3,7-9H

InChI key

GGNQRNBDZQJCCN-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 137 °C
Melting range (upper value): ≤ 140 °C
Identity (IR): passes test


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters