Skip to Content
Merck

Skip To

8.43546

Methyl palmitate

for synthesis

Synonym(s):

Methyl palmitate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 1,520.00
500 g
Please contact Customer Service for Availability
CZK 5,660.00

About This Item

Empirical Formula (Hill Notation):
C17H34O2
CAS Number:
Molecular Weight:
270.45
UNSPSC Code:
12352108
EC Index Number:
203-966-3
NACRES:
NA.22
MDL number:

CZK 1,520.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<0.01 hPa ( 20 °C)

Quality Segment

assay

≥97.0% (GC)

form

solid

bp

148 °C/3 hPa

mp

30 °C

transition temp

flash point 113 °C

storage temp.

2-30°C

SMILES string

O(C)C(=O)CCCCCCCCCCCCCCC

InChI

1S/C17H34O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h3-16H2,1-2H3

InChI key

FLIACVVOZYBSBS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 29 °C
Melting range (upper value): ≤ 31 °C
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR35385037476159
assay

≥97.0% (GC)

assay

-

assay

-

assay

≥98.5% (GC)

bp

148 °C/3 hPa

bp

185 °C/10 mmHg (lit.)

bp

185 °C/10 mmHg (lit.)

bp

185 °C/10 mmHg (lit.)

mp

30 °C

mp

32-35 °C (lit.)

mp

32-35 °C (lit.)

mp

32-35 °C (lit.)

transition temp

flash point 113 °C

transition temp

-

transition temp

-

transition temp

-

form

solid

form

-

form

-

form

-

vapor pressure

<0.01 hPa ( 20 °C)

vapor pressure

-

vapor pressure

-

vapor pressure

-


Still not finding the right product?

Explore all of our products under Methyl palmitate


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

235.4 °F - closed cup

flash_point_c

113.0 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters