Skip to Content
Merck

Skip To

8.41817

Tri(o-tolyl)-phosphine

for synthesis

Synonym(s):

Tri(o-tolyl)-phosphine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
CZK 1,460.00
10 g
Please contact Customer Service for Availability
CZK 4,320.00
CZK 3,240.00

About This Item

Empirical Formula (Hill Notation):
C21H21P
CAS Number:
Molecular Weight:
304.37
UNSPSC Code:
12352128
EC Index Number:
228-193-9
NACRES:
NA.22
MDL number:

CZK 1,460.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (HPLC)

form

solid

reaction suitability

reagent type: ligand

bp

238 °C/15 hPa

mp

124-127 °C

functional group

phosphine

storage temp.

2-30°C

SMILES string

P(c3c(cccc3)C)(c2c(cccc2)C)c1c(cccc1)C

InChI

1S/C21H21P/c1-16-10-4-7-13-19(16)22(20-14-8-5-11-17(20)2)21-15-9-6-12-18(21)3/h4-15H,1-3H3

InChI key

COIOYMYWGDAQPM-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 97.0 % (a/a)
Melting range (lower value): ≥ 124 °C
Melting range (upper value): ≤ 127 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
28782251885290572
assay

≥97.0% (HPLC)

assay

97%

assay

≥97.0% (HPLC)

assay

97%

reaction suitability

reagent type: ligand

reaction suitability

-

reaction suitability

-

reaction suitability

-

functional group

phosphine

functional group

phosphine

functional group

-

functional group

phosphine

form

solid

form

-

form

-

form

solid

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

200

storage temp.

2-30°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-


Still not finding the right product?

Explore all of our products under Tri(o-tolyl)-phosphine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters