Skip to Content
Merck

Skip To

8.41235

DL-Serine

for synthesis

Synonym(s):

DL-Serine, DL-2-Amino-3-hydroxypropionic acid, Ser

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 2,330.00

About This Item

Empirical Formula (Hill Notation):
C3H7NO3
CAS Number:
Molecular Weight:
105.09
UNSPSC Code:
12352209
EC Index Number:
206-130-6
NACRES:
NA.22
MDL number:

CZK 2,330.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

mp

239 °C (decomposition)

density

1.537 g/cm3

storage temp.

2-30°C

SMILES string

[N+H3]C(CO)C(=O)[O-]

InChI

1S/C3H7NO3/c4-2(1-5)3(6)7/h2,5H,1,4H2,(H,6,7)

InChI key

MTCFGRXMJLQNBG-UHFFFAOYSA-N

Analysis Note

Assay (HClO₄): ≥ 99.0 %
Melting range (lower value): ≥ 235 °C
Melting range (upper value): ≤ 240 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.149868.4156279710
form

powder

form

powder

form

powder

form

powder or crystals

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

mp

239 °C (decomposition)

mp

-

mp

303 °C

mp

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

density

1.537 g/cm3

density

1.293 g/cm3 at 18 °C

density

-

density

-


Still not finding the right product?

Explore all of our products under DL-Serine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters