Skip to Content
Merck

Skip To

8.22341

Triethanolamine

EMPLURA®

Synonym(s):

Triethanolamine, Tris(2-hydroxyethyl)amine, 2,2′,2′′-Trihydroxytriethylamine, TEA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 L
Please contact Customer Service for Availability
CZK 4,130.00
25 L
Please contact Customer Service for Availability
CZK 13,000.00

About This Item

Linear Formula:
N(CH2CH2OH)3
CAS Number:
Molecular Weight:
149.19
UNSPSC Code:
12352104
EC Index Number:
203-049-8
NACRES:
NA.21
MDL number:
Assay:
≥99.0% (GC)
Bp:
360 °C/1013 hPa (decomposition)
Vapor pressure:
<0.01 hPa ( 20 °C)

CZK 4,130.00


Please contact Customer Service for Availability
Packaging: PE canister

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

<0.01 hPa ( 20 °C)

Quality Segment

product line

EMPLURA®

assay

≥99.0% (GC)

form

liquid

autoignition temp.

325 °C (DIN 51794)

potency

>5000 mg/kg LD50, oral (Rat), >2000 mg/kg LD50, skin (Rabbit)

expl. lim.

3.6-7.2 % (v/v)

dilution

(low-cost alternative, for basic applications)

impurities

≤0.3% Water (Karl Fischer)

pH

10.5 (20 °C, 15 g/L in H2O)

bp

360 °C/1013 hPa (decomposition)

mp

21 °C

transition temp

flash point 190 °C

density

1.12 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

N(CCO)(CCO)CCO

InChI

1S/C6H15NO3/c8-4-1-7(2-5-9)3-6-10/h8-10H,1-6H2

InChI key

GSEJCLTVZPLZKY-UHFFFAOYSA-N

General description

EMPLURA® is our low-cost alternative to high-purity qualities. With EMPLURA we offer a range of solvents for a plurality of basic applications in non-regulated industries and for less demanding applications, preparative laboratory work and cleaning purposes. EMPLURA solvents provide adequate specifications with the most important parameters.

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.122 - 1.125
Water (K. F.): ≤ 0.30 %
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Legal Information

EMPLURA is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
33729902781.08372
assay

≥99.0% (GC)

assay

≥99% (GC)

assay

≥99.5% (GC)

assay

99.0-103.0% anhydrous basis (acidimetric)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

400

form

liquid

form

viscous liquid

form

liquid, viscous liquid

form

liquid

pH

10.5 (20 °C, 15 g/L in H2O)

pH

-

pH

10.5-11.5 (25 °C, 1 M in H2O)

pH

10.5 (20 °C, 15 g/L in H2O)

impurities

≤0.3% Water (Karl Fischer)

impurities

≤0.0001% heavy metals (as Pb), ≤0.1% ethanolamine, ≤0.1% water (Karl Fischer), ≤0.5% diethanolamine (GC)

impurities

insoluble matter, passes filter test, ≤0.1% ethanolamine (GC), ≤0.2% water, ≤0.3% diethanolamine (GC)

impurities

-

expl. lim.

3.6-7.2 % (v/v)

expl. lim.

8.5 %

expl. lim.

8.5 %

expl. lim.

3.6-7.2 % (v/v), 8.5 %


Still not finding the right product?

Explore all of our products under Triethanolamine


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

354.2 °F - closed cup

flash_point_c

179 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Est ce que la DEA est dosée dans la référence de TEA suivante : 8.22341

    1 answer
    1. DEA content is not tested for this product.

      Helpful?

Reviews

No rating value

Active Filters