Skip to Content
Merck
All Photos(1)

Key Documents

8.21199

Sigma-Aldrich

Triphenylantimony

for synthesis

Synonym(s):

Triphenylantimony, Triphenylstibine, Antimony triphenyl

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 2,070.00

CZK 2,070.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
CZK 2,070.00

About This Item

Linear Formula:
(C6H5)3Sb
CAS Number:
Molecular Weight:
353.07
MDL number:
UNSPSC Code:
12352103
EC Index Number:
210-037-6
NACRES:
NA.22

CZK 2,070.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (HPLC)

form

powder

bp

377 °C/1013 hPa

mp

52-54 °C

transition temp

flash point 113 °C

solubility

<0.001 g/L

bulk density

1490 kg/m3

storage temp.

2-30°C

SMILES string

[Sb](c3ccccc3)(c2ccccc2)c1ccccc1

InChI

1S/3C6H5.Sb/c3*1-2-4-6-5-3-1;/h3*1-5H;

InChI key

HVYVMSPIJIWUNA-UHFFFAOYSA-N

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 52 °C
Melting range (upper value): ≤ 54 °C
Identity (IR): passes test

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service