Skip to Content
Merck
All Photos(1)

Key Documents

8.21120

Sigma-Aldrich

2-Thiouracil

for synthesis

Synonym(s):

2-Thiouracil, 4-Hydroxy-2-mercaptopyrimidine

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 2,220.00
500 G
CZK 7,980.00
1 KG
CZK 11,700.00

CZK 2,220.00


Please contact Customer Service for Availability


Select a Size

Change View
100 G
CZK 2,220.00
500 G
CZK 7,980.00
1 KG
CZK 11,700.00

About This Item

Empirical Formula (Hill Notation):
C4H4N2OS
CAS Number:
Molecular Weight:
128.15
MDL number:
UNSPSC Code:
12352115
EC Index Number:
205-508-8
NACRES:
NA.22

CZK 2,220.00


Please contact Customer Service for Availability

Quality Level

Assay

≥98.0% (HPLC)

autoignition temp.

570 °C

mp

>300 °C

bulk density

600 kg/m3

storage temp.

2-30°C

SMILES string

Sc1nc(ccn1)O

InChI

1S/C4H4N2OS/c7-3-1-2-5-4(8)6-3/h1-2H,(H2,5,6,7,8)

InChI key

ZEMGGZBWXRYJHK-UHFFFAOYSA-N

Application

<ul>
<li><strong>Enhanced photocatalytic degradation using 2-Thiouracil:</strong> 2-Thiouracil is used obtained a carbonyl heterocycle-containing mesoporous carbon nitride photocatalyst (TCN) that demonstrated improved photocatalytic activity for environmental pollutant degradation, specifically for ciprofloxacin removal (Xu et al., 2023).</li>
</ul>

Analysis Note

Assay (HPLC, area%): ≥ 98.0 % (a/a)
Identity (IR): passes test

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service