Skip to Content
Merck

Skip To

8.20756

Malondialdehyde bis(dimethyl acetal)

for synthesis

Synonym(s):

Malondialdehyde bis(dimethyl acetal), 1,1,3,3-Tetramethoxypropane, Malondialdehyde tetramethyl acetal

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mL
Please contact Customer Service for Availability
CZK 688.00
250 mL
Please contact Customer Service for Availability
CZK 2,180.00

About This Item

Linear Formula:
(CH3O)2CHCH2CH(OCH3)2
CAS Number:
Molecular Weight:
164.20
UNSPSC Code:
12352112
EC Index Number:
203-037-2
NACRES:
NA.22
MDL number:
eCl@ss:
39021012
Form:
liquid

CZK 688.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

1.7 hPa ( 20 °C)

Quality Segment

form

liquid

potency

2440 mg/kg LD50, oral (Rat)

pH

7 (20 °C in H2O)

kinematic viscosity

1.75 cSt(20 °C)

bp

183 °C

transition temp

flash point 57 °C

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(C(OC)CC(OC)OC)C

InChI

1S/C7H16O4/c1-8-6(9-2)5-7(10-3)11-4/h6-7H,5H2,1-4H3

InChI key

XHTYQFMRBQUCPX-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.200028.0136663287
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

bp

183 °C

bp

63-65 °C/1013 hPa

bp

102 °C/1013 hPa

bp

-

density

0.99 g/cm3 at 20 °C

density

0.85 g/cm3 at 20 °C

density

0.83 g/cm3 at 20 °C

density

-

form

liquid

form

liquid

form

liquid

form

solid

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-8°C

transition temp

flash point 57 °C

transition temp

flash point -17 °C

transition temp

flash point -21 °C

transition temp

-


pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

134.6 °F - closed cup - DIN 51755 Part 1

flash_point_c

57 °C - closed cup - DIN 51755 Part 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters