Skip to Content
Merck

Skip To

8.20665

1,4-Butanesultone

for synthesis

Synonym(s):

1,4-Butanesultone, 4-Hydroxy-1-butanesulfonic acid δ-sultone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
50 mL
Please contact Customer Service for Availability
CZK 2,340.00
250 mL
Please contact Customer Service for Availability
CZK 7,070.00

About This Item

Empirical Formula (Hill Notation):
C4H8O3S
CAS Number:
Molecular Weight:
136.17
UNSPSC Code:
12352119
EC Index Number:
216-647-9
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Form:
liquid

CZK 2,340.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98.0% (GC)

form

liquid

potency

500 mg/kg LD50, oral (Rat)

bp

153 °C/19 hPa

mp

12.5-15.0 °C

transition temp

flash point 154 °C

solubility

54 g/L (decomposition)

density

1.34 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

[S]1(=O)(=O)OCCCC1

InChI

1S/C4H8O3S/c5-8(6)4-2-1-3-7-8/h1-4H2

InChI key

MHYFEEDKONKGEB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.334 - 1.336
Identity (IR): passes test
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.


Still not finding the right product?

Explore all of our products under 1,4-Butanesultone


signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

309.2 °F

flash_point_c

154 °C

pictograms

Health hazardExclamation mark


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters