Skip to Content
Merck
All Photos(1)

Key Documents

8.20433

Sigma-Aldrich

2,4-Dichlorobenzaldehyde

for synthesis

Synonym(s):

2,4-Dichlorobenzaldehyde

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 1,100.00
500 G
CZK 3,820.00

CZK 1,100.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
CZK 1,100.00
500 G
CZK 3,820.00

About This Item

Linear Formula:
2,4-(Cl)2C6H3CHO
CAS Number:
Molecular Weight:
175.01
MDL number:
UNSPSC Code:
12352114
EC Index Number:
212-861-1
NACRES:
NA.22

CZK 1,100.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

form

crystals

autoignition temp.

440 °C

potency

3470 mg/kg LD50, oral (Rat)

bp

233 °C/1013 hPa

mp

68-71 °C

transition temp

flash point 135 °C

bulk density

700 kg/m3

storage temp.

2-30°C

SMILES string

Clc1c(ccc(c1)Cl)C=O

InChI

1S/C7H4Cl2O/c8-6-2-1-5(4-10)7(9)3-6/h1-4H

InChI key

YSFBEAASFUWWHU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 68 °C
Melting range (upper value): ≤ 71 °C
Identity (IR): passes test

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service