Skip to Content
Merck

Skip To

8.20159

Bis(2-chloroethyl)ammonium chloride

for synthesis

Synonym(s):

Bis(2-chloroethyl)ammonium chloride, Bis(2-chloroethyl)amine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 g
Please contact Customer Service for Availability
CZK 1,040.00
CZK 780.00

About This Item

Linear Formula:
(ClCH2CH2)2NH2Cl
CAS Number:
Molecular Weight:
178.49
UNSPSC Code:
12352107
EC Index Number:
212-479-5
NACRES:
NA.22
MDL number:
Form:
powder

CZK 780.00

List PriceCZK 1,040.00Save 25%

Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

potency

1150 mg/kg LD50, oral (Rat)

mp

212-215 °C (decomposition)

solubility

>100 g/L

storage temp.

2-30°C

SMILES string

[Cl-].ClCC[N+H2]CCCl

InChI

1S/C4H9Cl2N.ClH/c5-1-3-7-4-2-6;/h7H,1-4H2;1H

InChI key

YMDZDFSUDFLGMX-UHFFFAOYSA-N

Analysis Note

Assay (argentometric): ≥ 98.0 %
Melting range (lower value): ≥ 210 °C
Melting range (upper value): ≤ 215 °C
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.016788.41417D65607
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

solubility

>100 g/L

solubility

-

solubility

-

solubility

-

storage temp.

2-30°C

storage temp.

-

storage temp.

2-30°C

storage temp.

-

form

powder

form

liquid

form

powder

form

solid

mp

212-215 °C (decomposition)

mp

-51.9 °C

mp

182-184 °C (decomposition)

mp

105-107 °C (lit.)

potency

1150 mg/kg LD50, oral (Rat)

potency

-

potency

96 mg/kg LD50, oral (Rat), 1502 mg/kg LD50, skin (Rat)

potency

-


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 3

flash_point_f

No information available.

flash_point_c

No information available.



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters