Skip to Content
Merck
All Photos(1)

Key Documents

8.20117

Sigma-Aldrich

Azelaic acid

for synthesis

Synonym(s):

Azelaic acid

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CZK 1,610.00
100 G
CZK 3,360.00

CZK 1,610.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
CZK 1,610.00
100 G
CZK 3,360.00

About This Item

Empirical Formula (Hill Notation):
C9H16O4
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352106
EC Index Number:
204-669-1
NACRES:
NA.22

CZK 1,610.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

1.3 hPa ( 178.3 °C)

Quality Level

Assay

≥98.0% (GC)

form

solid

potency

>4000 mg/kg LD50, oral (Rat)
>10 g/kg LD50, skin (Rat)

bp

237 °C/20 hPa

mp

106-109 °C

transition temp

flash point 210 °C

storage temp.

2-30°C

SMILES string

[O-]C(=O)CCCCCCCC(=O)[O-].[H+].[H+]

InChI

1S/C9H16O4/c10-8(11)6-4-2-1-3-5-7-9(12)13/h1-7H2,(H,10,11)(H,12,13)

InChI key

BDJRBEYXGGNYIS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Melting range (lower value): ≥ 106 °C
Melting range (upper value): ≤ 109 °C
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

410.0 °F - closed cup

Flash Point(C)

210 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service