Skip to Content
Merck
All Photos(1)

Key Documents

8.20082

Sigma-Aldrich

Aminoiminomethanesulfinic acid

for synthesis

Synonym(s):

Aminoiminomethanesulfinic acid, FormamidinesulfInic acid, Thiourea dioxide

Sign Into View Organizational & Contract Pricing

Select a Size

250 G
CZK 1,890.00
1 KG
CZK 4,300.00

CZK 1,890.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
250 G
CZK 1,890.00
1 KG
CZK 4,300.00

About This Item

Linear Formula:
HN=C(NH2)SO2H
CAS Number:
Molecular Weight:
108.12
MDL number:
UNSPSC Code:
12352106
EC Index Number:
217-157-8
NACRES:
NA.22

CZK 1,890.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

crystals

potency

>2000 mg/kg LD50, skin (Rat)

reaction suitability

reagent type: reductant

pH

4 (20 °C, 10 g/L in H2O)

mp

123 °C (decomposition)

solubility

27 g/L

density

1.68 g/cm3 at 20 °C

bulk density

850 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(O)\C(=N\[H])\N

InChI

1S/CH4N2O2S/c2-1(3)6(4)5/h(H3,2,3)(H,4,5)

InChI key

FYOWZTWVYZOZSI-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Self-heat. 1 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Lungs, Respiratory system

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service