Skip to Content
Merck
All Photos(1)

Key Documents

8.18858

Sigma-Aldrich

Tetra-n-butylammonium hydrogen sulfate

for synthesis

Synonym(s):

Tetra-n-butylammonium hydrogen sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
CZK 2,370.00
500 G
CZK 7,000.00

CZK 2,370.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 G
CZK 2,370.00
500 G
CZK 7,000.00

About This Item

Empirical Formula (Hill Notation):
C16H37NO4S
CAS Number:
Molecular Weight:
339.53
MDL number:
UNSPSC Code:
12352107
EC Index Number:
251-068-5
NACRES:
NA.22

CZK 2,370.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (acidimetric)

form

crystals

pH

1-2 (20 °C, 100 g/L in H2O)

mp

168-174 °C

bulk density

650 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])[O-].[N+](CCCC)(CCCC)(CCCC)CCCC.[H+]

InChI

1S/C16H36N.H2O4S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-5(2,3)4/h5-16H2,1-4H3;(H2,1,2,3,4)/q+1;/p-1

InChI key

SHFJWMWCIHQNCP-UHFFFAOYSA-M

Application


  • Effective Oxidation Catalysis: It enhances the oxidation of benzylic and alkane C-H bonds under phase-transfer conditions, using Oxone as a terminal oxidant, indicating its role in improving oxidation reactions in organic chemistry (Cui et al., 2011).

  • Hydrogen Bonding Studies: The interaction of Tetra-n-butylammonium hydrogen sulfate with protic acids has been studied, providing insights into hydrogen bond-controlled adduct formation critical for molecular recognition processes in chemical engineering (Zakavi et al., 2010).

Analysis Note

Assay (acidimetric): ≥ 98.0 %
Melting range (lower value): ≥ 167 °C
Melting range (upper value): ≤ 173 °C
Identity (IR): passes test

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1

Storage Class Code

8A - Combustible, corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

348.1 °F - closed cup

Flash Point(C)

175.6 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service