Skip to Content
Merck

Skip To

8.18339

2-Methoxyphenol

for synthesis

Synonym(s):

2-Methoxyphenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
5 mL
Please contact Customer Service for Availability
CZK 756.00
250 mL
Please contact Customer Service for Availability
CZK 1,650.00
1 L
Please contact Customer Service for Availability
CZK 5,010.00

About This Item

Empirical Formula (Hill Notation):
C7H8O2
CAS Number:
Molecular Weight:
124.14
UNSPSC Code:
12352104
EC Index Number:
201-964-7
NACRES:
NA.22
MDL number:
Assay:
≥98.0% (GC)
Grade:
synthesis grade
Bp:
205 °C/1013 hPa
Vapor pressure:
0.1 hPa ( 25 °C)

CZK 756.00


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

synthesis grade

Quality Segment

vapor pressure

0.1 hPa ( 25 °C)

assay

≥98.0% (GC)

form

solid

autoignition temp.

750 °C

potency

520 mg/kg LD50, oral (Rat), 4600 mg/kg LD50, skin (Rabbit)

dilution

(for synthesis)

pH

5.4 (20 °C, 10 g/L in H2O)

bp

205 °C/1013 hPa

mp

28-32 °C

transition temp

flash point 82 °C

density

1.13 g/cm3 at 21 °C

storage temp.

2-30°C

SMILES string

O(C)c1c(cccc1)O

InChI

1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3

InChI key

LHGVFZTZFXWLCP-UHFFFAOYSA-N

Analysis Note

Identity (IR): passes test
Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.132 - 1.134
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
CRM40484PHR1136W253200
grade

synthesis grade

grade

certified reference material, TraceCERT®

grade

certified reference material, pharmaceutical secondary standard

grade

FG, Fragrance grade, Halal, Kosher, natural

assay

≥98.0% (GC)

assay

-

assay

-

assay

≥99%

Quality Level

200

Quality Level

300

Quality Level

300

Quality Level

300, 400

form

solid

form

-

form

-

form

liquid, solid

mp

28-32 °C

mp

-

mp

26-29 °C (lit.)

mp

26-29 °C (lit.)

vapor pressure

0.1 hPa ( 25 °C)

vapor pressure

-

vapor pressure

0.11 mmHg ( 25 °C)

vapor pressure

0.11 mmHg ( 25 °C)


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

194.0 °F - closed cup

flash_point_c

90 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters