Skip to Content
Merck
All Photos(1)

Key Documents

8.18068

Sigma-Aldrich

Hydrogen fluoride

(65% solution in pyridin) for synthesis

Synonym(s):

Hydrogen fluoride

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
CZK 5,820.00

CZK 5,820.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
CZK 5,820.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.22

CZK 5,820.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

liquid

pH

<7 (20 °C in H2O)

density

1.14 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[F-].n1ccccc1.[H+]

InChI

1S/C5H5N.FH/c1-2-4-6-5-3-1;/h1-5H;1H

InChI key

GRJJQCWNZGRKAU-UHFFFAOYSA-N

Analysis Note

Assay (acidimetric): 63.0 - 67.0 %

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

68.0 °F - closed cup

Flash Point(C)

20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service