Skip to Content
Merck

Skip To

8.18003

Silver trifluoroacetate

for synthesis

Synonym(s):

Silver trifluoroacetate, Trifluoroacetic acid silver salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Please contact Customer Service for Availability
CZK 18,200.00

About This Item

Empirical Formula (Hill Notation):
C2AgF3O2
CAS Number:
Molecular Weight:
220.88
UNSPSC Code:
12352103
EC Index Number:
221-004-0
NACRES:
NA.22
MDL number:

CZK 18,200.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder

reaction suitability

reagent type: oxidant

mp

257-260 °C

storage temp.

2-30°C

SMILES string

[Ag]OC(=O)C(F)(F)F

InChI

1S/C2HF3O2.Ag/c3-2(4,5)1(6)7;/h(H,6,7);/q;+1/p-1

InChI key

KZJPVUDYAMEDRM-UHFFFAOYSA-M

Analysis Note

Assay (from Ag): ≥ 98
Identity (IR-spectrum): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.143258.188818.01504
form

powder

form

powder

form

crystals

form

crystals

mp

257-260 °C

mp

356 °C

mp

70-73 °C

mp

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-30°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

reaction suitability

reagent type: oxidant

reaction suitability

-

reaction suitability

reagent type: oxidant

reaction suitability

-


Still not finding the right product?

Explore all of our products under Silver trifluoroacetate


pictograms

Skull and crossbonesEnvironment

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters