Skip to Content
Merck

Skip To

8.15103

Quinoline-2,4-dicarboxylic acid

for synthesis

Synonym(s):

Quinoline-2,4-dicarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 g
Please contact Customer Service for Availability
CZK 5,350.00

About This Item

Empirical Formula (Hill Notation):
C11H7NO4
CAS Number:
Molecular Weight:
217.18
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.22
Form:
solid

CZK 5,350.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

solid

mp

229 °C

storage temp.

2-30°C

SMILES string

n1c2c(c(cc1C(=O)O)C(=O)O)cccc2

InChI

1S/C11H7NO4/c13-10(14)7-5-9(11(15)16)12-8-4-2-1-3-6(7)8/h1-5H,(H,13,14)(H,15,16)

InChI key

JPGZDTDZZDLVHW-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.00614792748153281
Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

100

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

-

mp

229 °C

mp

248-255 °C

mp

265-270 °C

mp

>300 °C (lit.)

form

solid

form

powder

form

powder

form

-


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters