Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
25 g | Please contact Customer Service for Availability | CZK 3,300.00 |
About This Item
Empirical Formula (Hill Notation):
C4H6N2S
CAS Number:
Molecular Weight:
114.17
UNSPSC Code:
12352005
EC Index Number:
248-473-4
NACRES:
NA.22
MDL number:
Form:
solid
Quality Segment
form
solid
potency
1000 mg/kg LD50, oral (Rat)
bp
202 °C/1013 hPa
mp
61-64 °C
storage temp.
2-30°C
SMILES string
[s]1c(nnc1C)C
InChI
1S/C4H6N2S/c1-3-5-6-4(2)7-3/h1-2H3
InChI key
JXQGICFGPUAVLJ-UHFFFAOYSA-N
1 of 1
This Item | |||
|---|---|---|---|
| Quality Level 200 | Quality Level 300 | Quality Level 100 | Quality Level 100 |
| bp 202 °C/1013 hPa | bp - | bp - | bp - |
| form solid | form - | form powder | form powder |
| mp 61-64 °C | mp - | mp 188-191 °C (dec.) (lit.) | mp - |
| storage temp. 2-30°C | storage temp. -10 to -25°C | storage temp. - | storage temp. 2-8°C |
| potency 1000 mg/kg LD50, oral (Rat) | potency - | potency - | potency - |
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class
11 - Combustible Solids
wgk
WGK 3
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.




