Skip to Content
Merck

Skip To

8.14858

Alkylbenzyldimethylammonium chloride

(50% solution in water) for synthesis

Synonym(s):

Benzalkonium chloride (50% aqueous solution)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 L
Please contact Customer Service for Availability
CZK 1,690.00
5 L
Please contact Customer Service for Availability
CZK 6,650.00

About This Item

NACRES:
NA.22
UNSPSC Code:
12352107
Form:
liquid

CZK 1,690.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

potency

500-2000 mg/kg LD50, oral (Rat)

pH

5-7 (20 °C, 10 g/L in H2O)

mp

-5 °C

transition temp

flash point 160-170 °C

density

0.978-0.988 g/cm3 at 20 °C

storage temp.

2-30°C

Application

  • Preservation of Cultural Heritage: Alkylbenzyldimethylammonium chloride have efficacy against microorganisms and is used in protecting painted cultural heritage objects (Alexandrova et al., 2024).
  • Evaluation in Glaucoma Treatments: The efficacy and safety of benzalkonium chloride-preserved intraocular pressure-lowering eye drops were analyzed, considering their impact on conjunctival goblet cells and implications for glaucoma treatment strategies (Nagstrup, 2023).

Analysis Note

Assay (argentometric): 49.0 - 51.0 %
Density (d 20 °C/ 4 °C): 0.980 - 0.990
Due to its specific melting range the product may be solid, liquid, a solidified melt or a supercooled melt.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.140871.00326PHR1681
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

300

form

liquid

form

liquid

form

liquid

form

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

15-25°C

storage temp.

2-30°C

density

0.978-0.988 g/cm3 at 20 °C

density

0.89 g/cm3 at 20 °C

density

0.79 g/cm3 at 20 °C

density

-

mp

-5 °C

mp

-

mp

-

mp

-

transition temp

flash point 160-170 °C

transition temp

flash point 15 °C

transition temp

flash point 12 °C (2-Propanol)

transition temp

-


Still not finding the right product?


signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible, corrosive hazardous materials

wgk

WGK 2

flash_point_f

320.0 - 338.0 °F

flash_point_c

160 - 170 °C



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters