Skip to Content
Merck

Skip To

8.14765

o-Dichlorobenzene/Phenol (mixture 1:1)

for synthesis

Synonym(s):

Phenol-1,2-dichlorobenzene mixture

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
2.5 L
Please contact Customer Service for Availability
CZK 12,700.00

About This Item

NACRES:
NA.22
UNSPSC Code:
12352401
Form:
liquid

CZK 12,700.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

transition temp

flash point 79 °C

density

1.18 g/cm3 at 20 °C

storage temp.

2-30°C

Application

  • Room-temperature-phase-selective gelation for the removal of organic pollutants: This study presents a novel gelation approach using o-Dichlorobenzene/Phenol mixture for effectively removing pollutants like nitrobenzene and dye molecules from biphasic mixtures (J Zhang et al., 2019).
  • Robust catalytic systems for chlorobenzene destruction: A study that enhances the catalytic destruction of chlorobenzene through the use of o-Dichlorobenzene/Phenol, addressing environmental pollution issues (L Li et al., 2021).
  • Catalytic hydroxylation of benzene to phenol: The paper discusses the specific enhancement of catalytic activity by a dicopper core using a 1:1 mixture of o-Dichlorobenzene and Phenol to achieve selective hydroxylation of benzene to phenol (T Tsuji et al., 2017).
  • Enhanced separation performance in gas chromatography: This research explores the use of Triptycene-based stationary phases for improving the separation of o-Dichlorobenzene and Phenol, enhancing analytical methodologies in chemistry (J He et al., 2019).

Analysis Note

Assay of phenole (GC, weight%): 48.0 - 52.0 %
Water (K. F.): ≤ 200 ppm
Assay of dichloro benzene (GC, weight%): 48.0 - 52.0 %
Colourless liquid: passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
33514335228.14506
Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

200

storage temp.

2-30°C

storage temp.

-

storage temp.

-

storage temp.

2-30°C

density

1.18 g/cm3 at 20 °C

density

-

density

-

density

1.11 g/cm3 at 20 °C

form

liquid

form

liquid

form

liquid

form

liquid

transition temp

flash point 79 °C

transition temp

-

transition temp

-

transition temp

flash point 27 °C


signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 3

wgk

WGK 2

flash_point_f

174.2 °F

flash_point_c

79 °C

target_organs

Nervous system,Kidney,Liver,Skin, Respiratory system

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters