Skip to Content
Merck
All Photos(1)

Key Documents

8.14764

Sigma-Aldrich

2,4,5-Triamino-6-hydroxypyrimidine sulfate

for synthesis

Synonym(s):

2,4,5-Triamino-6-hydroxypyrimidine sulfate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
CZK 1,370.00

CZK 1,370.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 G
CZK 1,370.00

About This Item

Empirical Formula (Hill Notation):
C4H7N5O · xH2SO4
CAS Number:
Molecular Weight:
141.13 (free base basis)
MDL number:
UNSPSC Code:
12352107
EC Index Number:
254-393-0
NACRES:
NA.22

CZK 1,370.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

form

powder

potency

>3000 mg/kg LD50, oral (Rat)

pH

2-3 (20 °C, 2 g/L in H2O)

mp

350 °C

solubility

2 g/L

bulk density

480 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])O.Nc1nc(c(c(n1)[O-])N)N

InChI

1S/C4H7N5O.H2O4S/c5-1-2(6)8-4(7)9-3(1)10;1-5(2,3)4/h5H2,(H5,6,7,8,9,10);(H2,1,2,3,4)/p-2

InChI key

RSKNEEODWFLVFF-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service