Skip to Content
Merck
All Photos(2)

Key Documents

8.14635

Sigma-Aldrich

2-(2-Ethoxyethoxy)ethyl acetate

for synthesis

Synonym(s):

2-(2-Ethoxyethoxy)ethyl acetate, Ethyl carbitol acetate, Diethylene glycol monoethyl ether acetate, Carbitolacetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
CZK 2,170.00

CZK 2,170.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 L
CZK 2,170.00

About This Item

Empirical Formula (Hill Notation):
C8H16O4
CAS Number:
Molecular Weight:
176.21
MDL number:
UNSPSC Code:
12352108
EC Index Number:
203-940-1
NACRES:
NA.22

CZK 2,170.00


Please contact Customer Service for Availability

Request a Bulk Order

vapor pressure

0.3 hPa ( 20 °C)

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

310 °C

potency

11000 mg/kg LD50, oral (Rat)
15251 mg/kg LD50, skin (Rabbit)

expl. lim.

1.5 % (v/v)

bp

217-218 °C

mp

-25 °C

transition temp

flash point 100 °C

density

1.01 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCOCC)CCOC(=O)C

InChI

1S/C8H16O4/c1-3-10-4-5-11-6-7-12-8(2)9/h3-7H2,1-2H3

InChI key

FPZWZCWUIYYYBU-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.006 - 1.011
Identity (IR): passes test

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

208.4 °F - closed cup

Flash Point(C)

98 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service