Skip to Content
Merck

Skip To

8.14546

DL-Limonene

(mixture of D- and L-form ~1:1) for synthesis

Synonym(s):

DL-Limonene, DL-p-Mentha-1,8-diene, 4-Isopropyl-1-methylcyclohexene

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 L
Please contact Customer Service for Availability
CZK 1,680.00

About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
UNSPSC Code:
12352002
EC Index Number:
205-341-0
NACRES:
NA.22
MDL number:
Assay:
≥95.0% (GC)
Form:
liquid

CZK 1,680.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

2.1 hPa ( 20 °C)

Quality Segment

assay

≥95.0% (GC)

form

liquid

autoignition temp.

255 °C

potency

5300 mg/kg LD50, oral (Rat)

expl. lim.

0.7-6.1 % (v/v)

bp

178 °C/1013 hPa

mp

-89 °C

transition temp

flash point 43 °C

density

0.84 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

C1(CCC(=CC1)C)C(=C)C

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,10H,1,5-7H2,2-3H3

InChI key

XMGQYMWWDOXHJM-UHFFFAOYSA-N

Application

  • Antimicrobial and anti-inflammatory activities of essential oils: A study evaluates the antimicrobial and anti-inflammatory activities of essential oils from Juniperus species, with a focus on compounds including DL-limonene, suggesting its utility in ethnopharmacology and as a green solvent in medicinal plant research (Darwish et al., 2020).

Analysis Note

Assay (GC, area%): ≥ 95.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.841 - 0.845
Identity (IR): passes test

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.1840718316417763
assay

≥95.0% (GC)

assay

≥94.0% (GC)

assay

97%

assay

≥97.0% (sum of isomers, GC)

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

100

form

liquid

form

liquid

form

liquid

form

liquid

bp

178 °C/1013 hPa

bp

177-178 °C/1013 hPa

bp

176-177 °C (lit.)

bp

-

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

-

storage temp.

2-8°C

density

0.84 g/cm3 at 20 °C

density

0.84 g/cm3 at 20 °C

density

0.842 g/mL at 20 °C (lit.)

density

0.930 g/mL at 20 °C (lit.)


signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

109.4 °F - closed cup

flash_point_c

43 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters