Skip to Content
Merck
All Photos(1)

Key Documents

8.14500

Sigma-Aldrich

Isoamyl benzoate

for synthesis

Synonym(s):

Isoamyl benzoate, Benzoic acid isoamyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

5 ML
CZK 562.00
250 ML
CZK 1,500.00

CZK 562.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 ML
CZK 562.00
250 ML
CZK 1,500.00

About This Item

Empirical Formula (Hill Notation):
C12H16O2
CAS Number:
Molecular Weight:
192.25
MDL number:
UNSPSC Code:
12352108
EC Index Number:
202-334-4
NACRES:
NA.22

CZK 562.00


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥99.0% (GC)

form

liquid

potency

6330 mg/kg LD50, oral (Rat)

bp

260 °C

transition temp

flash point 89 °C

density

0.99 g/cm3 at 20 °C

storage temp.

2-30°C

SMILES string

O(CCC(C)C)C(=O)c1ccccc1

InChI

1S/C12H16O2/c1-10(2)8-9-14-12(13)11-6-4-3-5-7-11/h3-7,10H,8-9H2,1-2H3

InChI key

MLLAPOCBLWUFAP-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.988 - 0.990
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service