Skip to Content
Merck
All Photos(1)

Key Documents

8.14410

Sigma-Aldrich

Azetidine

for synthesis

Synonym(s):

Azetidine, Trimethylene imine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7N
CAS Number:
Molecular Weight:
57.09
MDL number:
UNSPSC Code:
12352005
EC Index Number:
207-963-8

vapor pressure

237 hPa ( 25 °C)

Quality Level

Assay

≥99.0% (GC)

transition temp

flash point -20 °C

density

0.85 g/cm3 at 20 °C

InChI

1S/C3H7N/c1-2-4-3-1/h4H,1-3H2

InChI key

HONIICLYMWZJFZ-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.844 - 0.847
Identity (IR): passes test

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service