Skip to Content
Merck

Skip To

8.10953

Kryptofix® 22

for synthesis

Synonym(s):

Kryptofix® 22, 1,7,10,16-Tetraoxa-4,13-diazacyclooctadecane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Please contact Customer Service for Availability
CZK 1,820.00
5 g
Please contact Customer Service for Availability
CZK 6,140.00

About This Item

Empirical Formula (Hill Notation):
C12H26N2O4
CAS Number:
Molecular Weight:
262.35
UNSPSC Code:
12352005
EC Index Number:
245-965-0
NACRES:
NA.22
MDL number:
Assay:
99%
Form:
powder

CZK 1,820.00


Please contact Customer Service for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

99%

form

powder

potency

3400 mg/kg LD50, oral (Rat)

reaction suitability

core: cryptand

mp

112-115 °C

storage temp.

2-30°C

SMILES string

[N+H2]1CCOCCOCC[N+H2]CCOCCOCC1

InChI

1S/C12H26N2O4/c1-5-15-9-10-17-7-3-14-4-8-18-12-11-16-6-2-13-1/h13-14H,1-12H2/p+2

InChI key

NLMDJJTUQPXZFG-UHFFFAOYSA-P

Application

Kryptofix® 22 for synthesis. CAS 23978-55-4, molar mass 262.35 g/mol.

Analysis Note

Purity (GC): ≥ 99
Identity (IR-spectrum): passes test
Melting point: 112 - 115

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.106478.106468.11689
assay

99%

assay

99%

assay

98%

assay

-

reaction suitability

core: cryptand

reaction suitability

core: cryptand

reaction suitability

core: cryptand, reagent type: reductant

reaction suitability

core: crown ether, core: cryptand

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

viscous liquid

form

powder

mp

112-115 °C

mp

70-73 °C

mp

-

mp

72-76 °C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-30°C


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters